• USA Home
  • B3895 - (2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

EMAIL THIS PAGE TO A FRIEND
B3895 Sigma-Aldrich

(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

>99% (TLC)

Synonym: Boc-(2S,3R)-3-amino-2-hydroxy-4-phenylbutyric acid

  • CAS Number 62023-65-8

  • Empirical Formula (Hill Notation) C15H21NO5

  • Molecular Weight 295.33

  •  Beilstein/REAXYS Number 5909790

  •  MDL number MFCD00057841

Purchase

Properties

assay   >99% (TLC)
storage temp.   −20°C
SMILES string   CC(C)(C)OC(=O)N[C@H](Cc1ccccc1)[C@H](O)C(O)=O
InChI   1S/C15H21NO5/c1-15(2,3)21-14(20)16-11(12(17)13(18)19)9-10-7-5-4-6-8-10/h4-8,11-12,17H,9H2,1-3H3,(H,16,20)(H,18,19)/t11-,12+/m1/s1
InChI key   BHTRKISIDQZUQX-NEPJUHHUSA-N

Description

Application

Novel protected amino acid for the preparation of bestatin and analogs.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?