• USA Home
  • B4028 - Boc-β-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin hydrochloride

EMAIL THIS PAGE TO A FRIEND
B4028 Sigma-Aldrich

Boc-β-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin hydrochloride

~95%

  • CAS Number 113866-00-5

  • Empirical Formula (Hill Notation) C37H47N7O9 · HCl

  • Molecular Weight 770.27

  •  MDL number MFCD00080215

  •  PubChem Substance ID 24891749

Purchase

Properties

assay   ~95%
storage temp.   −20°C
SMILES string   CC1=CC(=O)Oc2cc(NC(=O)C(CCCNC(N)=N)NC(=O)C3CCCN3C(=O)C(CC(=O)OCc4ccccc4)NC(=O)OC(C)(C)C)ccc12

Description

Substrates

Substrate for α-thrombin

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?