• USA Home
  • B4125 - Nα-Benzoyl-DL-arginine

EMAIL THIS PAGE TO A FRIEND
B4125 Sigma

Nα-Benzoyl-DL-arginine

  • CAS Number 6453-58-3

  • Empirical Formula (Hill Notation) C13H18N4O3

  • Molecular Weight 278.31

  •  MDL number MFCD00063011

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?