EMAIL THIS PAGE TO A FRIEND
B4138 Sigma-Aldrich

Boc-D-Met-OH

Synonym: Boc-D-methionine

  • CAS Number 5241-66-7

  • Empirical Formula (Hill Notation) C10H19NO4S

  • Molecular Weight 249.33

  •  Beilstein/REAXYS Number 4294122

  •  EC Number 226-043-7

  •  MDL number MFCD00038256

Purchase

Properties

InChI Key   IMUSLIHRIYOHEV-SSDOTTSWSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15132 Boc-D-Met-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?