EMAIL THIS PAGE TO A FRIEND
B4379 Sigma-Aldrich

Boc-Thr(Bzl)-OH

Synonym: N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine, Boc-O-benzyl-L-threonine

  • CAS Number 15260-10-3

  • Linear Formula CH3CH(OCH2C6H5)CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 309.36

  •  Beilstein/REAXYS Number 3065591

  •  EC Number 239-304-5

  •  MDL number MFCD00066062

  •  PubChem Substance ID 329773233

Purchase

Properties

mp   112-114 °C (lit.)
storage temp.   −20°C
SMILES string   C[C@@H](OCc1ccccc1)[C@H](NC(=O)OC(C)(C)C)C(O)=O
InChI   1S/C16H23NO5/c1-11(21-10-12-8-6-5-7-9-12)13(14(18)19)17-15(20)22-16(2,3)4/h5-9,11,13H,10H2,1-4H3,(H,17,20)(H,18,19)/t11-,13+/m1/s1
InChI key   CTXPLTPDOISPTE-YPMHNXCESA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15405 Boc-Thr(Bzl)-OH, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?