• USA Home
  • B4506 - N-Benzoyl-DL-phenylalanine β-naphthol AS-BI ester

EMAIL THIS PAGE TO A FRIEND
B4506 Sigma-Aldrich

N-Benzoyl-DL-phenylalanine β-naphthol AS-BI ester

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?