• USA Home
  • B4631 - New Methylene Blue N zinc chloride double salt

EMAIL THIS PAGE TO A FRIEND
B4631 Sigma

New Methylene Blue N zinc chloride double salt

Synonym: Basic Blue 24, Methylene Blue N

  • CAS Number 6586-05-6

  • Linear Formula C18H22N3SCl · 1/2ZnCl2

  • Molecular Weight 416.05

  •  Colour Index Number 52030

  •  EC Number 229-516-6

  •  MDL number MFCD00078135

Purchase

Properties

composition   Dye content, ~90%
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

202096 New Methylene Blue N zinc chloride double salt, Dye content 90 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?