EMAIL THIS PAGE TO A FRIEND
B4755 Sigma-Aldrich

1-Docosanol

~98%

Synonym: Behenyl alcohol

  • CAS Number 661-19-8

  • Linear Formula CH3(CH2)21OH

  • Molecular Weight 326.60

  •  Beilstein/REAXYS Number 1770470

  •  EC Number 211-546-6

  •  MDL number MFCD00002939

Purchase

Properties

assay   ~98%
bp   180 °C/0.22 mmHg (lit.)
mp   65-72 °C (lit.)
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCCCCCO
InChI   1S/C22H46O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3
InChI key   NOPFSRXAKWQILS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

169102 1-Docosanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?