• USA Home
  • B4782 - 5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt

EMAIL THIS PAGE TO A FRIEND
B4782 Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt

≥98% (TLC)

Synonym: BC-Indicator, X-GlcA, X-glucuronide

  • CAS Number 129541-41-9

  • Empirical Formula (Hill Notation) C14H12BrClNNaO7

  • Molecular Weight 444.59

  •  MDL number MFCD00135782

  •  PubChem Substance ID 24891703

Purchase

Properties

Related Categories Chromogenic, Glucuronide, Glycobiology, Inhibitors and Substrates, Molecular Biology,
assay   ≥98% (TLC)
storage temp.   −20°C
SMILES string   [Na+].O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C([O-])=O)Oc2c[nH]c3ccc(Br)c(Cl)c23
InChI   1S/C14H13BrClNO7.Na/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;/h1-3,9-12,14,17-20H,(H,21,22);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1
InChI key   IBLSVGDGSKUDCT-CYRSAHDMSA-M

Description

Application

Chromogenic substrate for β-glucuronidase (GUS) gene detection.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B5285 5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?