EMAIL THIS PAGE TO A FRIEND
B5391 Sigma

9-Bromo-1-nonanol

~95%

Synonym: Nonamethylene bromohydrin

  • CAS Number 55362-80-6

  • Linear Formula Br(CH2)9OH

  • Molecular Weight 223.15

  •  Beilstein Registry Number 1737525

  •  MDL number MFCD00063348

Purchase

Properties

InChI Key   USJDOLXCPFASNV-UHFFFAOYSA-N
assay   ~95%
bp   125-126 °C/2 mmHg(lit.)
mp   33-35 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?