EMAIL THIS PAGE TO A FRIEND
B5501 Sigma-Aldrich

Boc-Asn-ONp

95-98%

Synonym: Nα-Boc-L-asparagine 4-nitrophenyl ester

  • CAS Number 4587-33-1

  • Empirical Formula (Hill Notation) C15H19N3O7

  • Molecular Weight 353.33

  •  Beilstein/REAXYS Number 1895460

  •  MDL number MFCD00056109

Purchase

Properties

assay   95-98%
storage temp.   −20°C
SMILES string   CC(C)(C)OC(=O)N[C@@H](CC(N)=O)C(=O)Oc1ccc(cc1)[N+]([O-])=O
InChI   1S/C15H19N3O7/c1-15(2,3)25-14(21)17-11(8-12(16)19)13(20)24-10-6-4-9(5-7-10)18(22)23/h4-7,11H,8H2,1-3H3,(H2,16,19)(H,17,21)/t11-/m0/s1
InChI key   IAPXDJMULQXGDD-NSHDSACASA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?