EMAIL THIS PAGE TO A FRIEND
B6505 Sigma

Boc-His(Tos)-OH

Synonym: Nα-Boc-N(im)-tosyl-L-histidine

  • CAS Number 35899-43-5

  • Empirical Formula (Hill Notation) C18H23N3O6S

  • Molecular Weight 409.46

  •  Beilstein Registry Number 769957

  •  MDL number MFCD00065967

Purchase

Properties

InChI Key   DCLJSEPKYJSEHW-HNNXBMFYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15559 Boc-His(Tos)-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?