• USA Home
  • B6762 - (R)-(+)-1,1′-Bi(2-naphthol)

EMAIL THIS PAGE TO A FRIEND
B6762 Sigma-Aldrich

(R)-(+)-1,1′-Bi(2-naphthol)

Synonym: (+)-2,2′-Dihydroxy-1,1′-dinaphthyl, (R)-(+)-1,1′-Binaphthalene-2,2′-diol, (R)-BINOL

  • CAS Number 18531-94-7

  • Linear Formula HOC10H6C10H6OH

  • Molecular Weight 286.32

  •  Beilstein/REAXYS Number 3616837

  •  MDL number MFCD00004068

Purchase

Properties

InChI Key   PPTXVXKCQZKFBN-UHFFFAOYSA-N
mp   208-210 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
DU3106100

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?