EMAIL THIS PAGE TO A FRIEND
B6776 Sigma-Aldrich

Bergenin monohydrate

  • CAS Number 108032-11-7

  • Empirical Formula (Hill Notation) C14H16O9 · H2O

  • Molecular Weight 346.29

  •  MDL number MFCD00167248

  •  PubChem Substance ID 24891822

Purchase

Properties

Related Categories Cell Biology, Nutrition Research, Other Biochemical, Phytochemicals by Chemical Classification
InChI Key   QCWSXSAFDSGKAT-YOKSUNLASA-N
mp   237-240 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?