• USA Home
  • B6887 - (S)-(−)-1,1′-Bi(2-naphthol)

EMAIL THIS PAGE TO A FRIEND
B6887 Sigma

(S)-(−)-1,1′-Bi(2-naphthol)

Synonym: (−)-2,2′-Dihydroxy-1,1′-dinaphthyl, (S)-(−)-1,1′-Binaphthalene-2,2′-diol, (S)-BINOL

  • CAS Number 18531-99-2

  • Linear Formula HOC10H6C10H6OH

  • Molecular Weight 286.32

  •  Beilstein/REAXYS Number 4296068

  •  MDL number MFCD00004068

Purchase

Properties

InChI Key   PPTXVXKCQZKFBN-UHFFFAOYSA-N
mp   208-210 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
DU3106100

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

246956 (S)-(−)-1,1′-Bi(2-naphthol), 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?