EMAIL THIS PAGE TO A FRIEND
B6905 Sigma-Aldrich

Boc-D-Chg-OH

Synonym: Boc-D-α-cyclohexylglycine

  • CAS Number 70491-05-3

  • Empirical Formula (Hill Notation) C13H23NO4

  • Molecular Weight 257.33

  •  Beilstein/REAXYS Number 7689661

  •  MDL number MFCD00133629

Purchase

Properties

InChI Key   QSUXZIPXYDQFCX-SNVBAGLBSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15089 Boc-D-Chg-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?