EMAIL THIS PAGE TO A FRIEND
B7751 Sigma

Boc-Arg(NO2)-OH

~99% (TLC)

Synonym: Nα-Boc-Nω-nitro-L-arginine

  • CAS Number 2188-18-3

  • Linear Formula HN=C(NHNO2)NH(CH2)3CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 319.31

  •  Beilstein Registry Number 2015163

  •  MDL number MFCD00065556

Purchase

Properties

InChI Key   OZSSOVRIEPAIMP-ZETCQYMHSA-N
assay   ~99% (TLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15470 Boc-Arg(NO2)-OH, ≥98.5% (calc. based on dry substance, T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?