• USA Home
  • B7882 - Z-Lys(Boc)-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
B7882 Sigma-Aldrich

Z-Lys(Boc)-OH (dicyclohexylammonium) salt

Synonym: Nα-Z-Nε-Boc-L-lysine (dicyclohexylammonium) salt, Nε-Boc-Nα-Z-L-lysine (dicyclohexylammonium) salt

  • CAS Number 2212-76-2

  • Empirical Formula (Hill Notation) C19H28N2O6 · C12H23N

  • Molecular Weight 561.75

  •  Beilstein/REAXYS Number 4650895

  •  EC Number 218-663-1

  •  MDL number MFCD00038269

Purchase

Properties

storage temp.   −20°C
SMILES string   C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCc3ccccc3)C(O)=O
InChI   1S/C19H28N2O6.C12H23N/c1-19(2,3)27-17(24)20-12-8-7-11-15(16(22)23)21-18(25)26-13-14-9-5-4-6-10-14;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h4-6,9-10,15H,7-8,11-13H2,1-3H3,(H,20,24)(H,21,25)(H,22,23);11-13H,1-10H2/t15-;/m0./s1
InChI key   VTDLJMATIHSOTR-RSAXXLAASA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15550 Z-Lys(Boc)-OH (dicyclohexylammonium) salt, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?