• USA Home
  • B8000 - S-Benzyl-L-cysteine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
B8000 Sigma-Aldrich

S-Benzyl-L-cysteine methyl ester hydrochloride

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?