• USA Home
  • B8133 - 1,1′-Dibenzyl-4,4′-bipyridinium dichloride

EMAIL THIS PAGE TO A FRIEND
B8133 Sigma-Aldrich

1,1′-Dibenzyl-4,4′-bipyridinium dichloride

Synonym: Benzyl viologen dichloride

  • CAS Number 1102-19-8

  • Empirical Formula (Hill Notation) C24H22Cl2N2

  • Molecular Weight 409.35

  •  Beilstein/REAXYS Number 3811585

  •  EC Number 214-158-5

  •  MDL number MFCD00011991

Purchase
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DW1930000

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?