EMAIL THIS PAGE TO A FRIEND
B8513 Sigma-Aldrich

Boc-D-Leu-OH

Synonym: Boc-D-leucine

  • CAS Number 16937-99-8

  • Empirical Formula (Hill Notation) C11H21NO4

  • Molecular Weight 231.29

  •  Beilstein/REAXYS Number 3657826

  •  MDL number MFCD00038294

Purchase

Properties

InChI Key   MDXGYYOJGPFFJL-MRVPVSSYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

15129 Boc-D-Leu-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?