EMAIL THIS PAGE TO A FRIEND
B9004 Sigma-Aldrich

Boc-Met-OH

Synonym: N-(tert-Butoxycarbonyl)-L-methionine, Boc-L-methionine

  • CAS Number 2488-15-5

  • Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 249.33

  •  Beilstein/REAXYS Number 1727869

  •  EC Number 219-639-3

  •  MDL number MFCD00065586

Purchase

Properties

InChI Key   IMUSLIHRIYOHEV-ZETCQYMHSA-N
mp   47-50 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?