EMAIL THIS PAGE TO A FRIEND
B9501 Sigma-Aldrich

Boc-Val-OH

Synonym: (S)-2-(Boc-amino)-3-methylbutyric acid, N-(tert-Butoxycarbonyl)-L-valine, Boc-L-valine

  • CAS Number 13734-41-3

  • Linear Formula (CH3)2CHCH(COOH)NHCOOC(CH3)3

  • Molecular Weight 217.26

  •  Beilstein/REAXYS Number 1711290

  •  EC Number 237-307-6

  •  MDL number MFCD00065605

  •  PubChem Substance ID 329773320

Purchase

Properties

mp   77-80 °C (lit.)
storage temp.   −20°C
SMILES string   CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
InChI   1S/C10H19NO4/c1-6(2)7(8(12)13)11-9(14)15-10(3,4)5/h6-7H,1-5H3,(H,11,14)(H,12,13)/t7-/m0/s1
InChI key   SZXBQTSZISFIAO-ZETCQYMHSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

15528 Boc-Val-OH, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?