• USA Home
  • B9525 - Butirosin A from Bacillus vitellinus

EMAIL THIS PAGE TO A FRIEND
B9525 Sigma-Aldrich

Butirosin A from Bacillus vitellinus

~95%

  • CAS Number 34291-02-6

  • Empirical Formula (Hill Notation) C21H41N5O12

  • Molecular Weight 555.58

  •  MDL number MFCD00078992

  •  PubChem Substance ID 24892143

Purchase

Properties

assay   ~95%
Mode of action   protein synthesis | interferes
storage temp.   2-8°C

Description

General description

Chemical structure: aminoglycoside

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WK2302000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?