EMAIL THIS PAGE TO A FRIEND
C0502 Sigma

Z-Gly-Pro

Synonym: Z-Glycyl-L-proline

  • CAS Number 1160-54-9

  • Empirical Formula (Hill Notation) C15H18N2O5

  • Molecular Weight 306.31

  •  Beilstein Registry Number 94536

  •  EC Number 214-598-8

  •  MDL number MFCD00037341

Purchase

Properties

InChI Key   ZTUKZKYDJMGJDC-LBPRGKRZSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

96270 Z-Gly-Pro, ≥99.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?