EMAIL THIS PAGE TO A FRIEND
C0533 Sigma

Z-D-Lys(Boc)-OH

Synonym: Nα-Cbz-Nε-Boc-D-Lysine, Nα-Z-Nε-Boc-D-lysine

  • CAS Number 66845-42-9

  • Empirical Formula (Hill Notation) C19H28N2O6

  • Molecular Weight 380.44

  •  Beilstein Registry Number 5310107

  •  MDL number MFCD00065699

Purchase

Properties

InChI Key   DYSBKEOCHROEGX-OAHLLOKOSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

96019 Z-D-Lys(Boc)-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?