• USA Home
  • C0656 - (1S)-(+)-Camphorquinone

EMAIL THIS PAGE TO A FRIEND
C0656 Sigma

(1S)-(+)-Camphorquinone

Synonym: (1S)-(+)-Bornanedione

  • CAS Number 2767-84-2

  • Empirical Formula (Hill Notation) C10H14O2

  • Molecular Weight 166.22

  •  Beilstein Registry Number 2613999

  •  EC Number 220-446-1

  •  MDL number MFCD00064159

Purchase

Properties

InChI Key   VNQXSTWCDUXYEZ-QUBYGPBYSA-N
mp   197-201 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

272078 (1S)-(+)-Camphorquinone, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?