• USA Home
  • C0878 - N-Chloroacetylglycine

EMAIL THIS PAGE TO A FRIEND
C0878 Sigma-Aldrich

N-Chloroacetylglycine

  • CAS Number 6319-96-6

  • Linear Formula C4H6NO3Cl

  • Molecular Weight 151.55

  •  MDL number MFCD00021733

  •  eCl@ss 32160406

  •  PubChem Substance ID 24892280

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   DLDTUYIGYMNERN-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?