• USA Home
  • C0909 - Sodium citrate tribasic dihydrate

EMAIL THIS PAGE TO A FRIEND
C0909 Sigma-Aldrich

Sodium citrate tribasic dihydrate

suitable for amino acid analysis, ≥98% (titration), powder

Synonym: Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

  • CAS Number 6132-04-3

  • Linear Formula HOC(COONa)(CH2COONa)2 · 2H2O

  • Molecular Weight 294.10

  •  Beilstein/REAXYS Number 6104939

  •  EC Number 200-675-3

  •  MDL number MFCD00150031

  •  PubChem Substance ID 24892286

Purchase

Properties

Related Categories Acids, Acids & Bases, Biochemicals and Reagents, Biological Buffers, Buffers A to Z,
InChI Key   NLJMYIDDQXHKNR-UHFFFAOYSA-K
assay   ≥98% (titration)
form   powder
color   white
mp   >300 °C (lit.)
solubility   H2O: 100 mg/mL
suitability   suitable for amino acid analysis
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?