EMAIL THIS PAGE TO A FRIEND
C1127 Sigma-Aldrich

Z-Hyp-OH

≥98% (TLC)

Synonym: N-Cbz-Hydroxy-L-proline, Z-L-4-Hydroxyproline

  • CAS Number 13504-85-3

  • Empirical Formula (Hill Notation) C13H15NO5

  • Molecular Weight 265.26

  •  Beilstein/REAXYS Number 90295

  •  EC Number 236-831-2

  •  MDL number MFCD00037329

Purchase

Properties

assay   ≥98% (TLC)
storage temp.   −20°C
SMILES string   O[C@@H]1C[C@H](N(C1)C(=O)OCc2ccccc2)C(O)=O
InChI   1S/C13H15NO5/c15-10-6-11(12(16)17)14(7-10)13(18)19-8-9-4-2-1-3-5-9/h1-5,10-11,15H,6-8H2,(H,16,17)/t10-,11+/m1/s1
InChI key   WWVCWLBEARZMAH-MNOVXSKESA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

96310 Z-Hyp-OH, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?