• USA Home
  • C1395 - (+)-Camphor-10-sulfonic acid

EMAIL THIS PAGE TO A FRIEND
C1395 Sigma-Aldrich

(+)-Camphor-10-sulfonic acid

Synonym: (+)-β-Camphorsulfonic acid, (1S)-(+)-Camphor-10-sulfonic acid

  • CAS Number 3144-16-9

  • Empirical Formula (Hill Notation) C10H16O4S

  • Molecular Weight 232.30

  •  Beilstein/REAXYS Number 2809675

  •  EC Number 221-554-1

  •  MDL number MFCD00064157

Purchase

Properties

mp   ~200 °C (dec.) (lit.)
storage temp.   2-8°C
SMILES string   [H][C@@]12CC[C@@](CS(O)(=O)=O)(C(=O)C1)C2(C)C
InChI   1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m1/s1
InChI key   MIOPJNTWMNEORI-GMSGAONNSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2585PSN2 8 / PGIII
WGK Germany 
3
RTECS 
ED1550000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C2107 (1S)-(+)-10-Camphorsulfonic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?