EMAIL THIS PAGE TO A FRIEND
C140 Sigma

4′-Chlorodiazepam

Synonym: 7-Chloro-5-(4-chlorophenyl)-1,3-dihydro-1-methyl-2H-1,4-benzodiazepin-2-one, Ro 5-4864, Ro5-4864

  • CAS Number 14439-61-3

  • Empirical Formula (Hill Notation) C16H12Cl2N2O

  • Molecular Weight 319.19

  •  Beilstein Registry Number 685202

  •  MDL number MFCD00043406

Purchase

Properties

InChI Key   PUMYFTJOWAJIKF-UHFFFAOYSA-N
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DF0500000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C5174 4′-Chlorodiazepam, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?