• USA Home
  • C1503 - N-Chloroacetyl-L-phenylalanine

EMAIL THIS PAGE TO A FRIEND
C1503 Sigma-Aldrich

N-Chloroacetyl-L-phenylalanine

  • CAS Number 721-65-3

  • Empirical Formula (Hill Notation) C11H12ClNO3

  • Molecular Weight 241.67

  •  MDL number MFCD00063160

  •  eCl@ss 32160406

  •  PubChem Substance ID 24892416

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
form   solid
application(s)   ligand binding assay: suitable
storage temp.   −20°C
SMILES string   OC(=O)[C@H](Cc1ccccc1)NC(=O)CCl
InChI   1S/C11H12ClNO3/c12-7-10(14)13-9(11(15)16)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,14)(H,15,16)/t9-/m0/s1
InChI key   FUHGSOAURCZWCC-VIFPVBQESA-N

Description

Biochem/physiol Actions

N-Chloroacetyl-L-phenylalanine is used as a substrate to identify, differentiate and characterize L-amino acid acylases such as A933 acylase. N-Chloroacetyl-L-phenylalanine may be a donor molecule in some chloroacetylation reactions.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?