• USA Home
  • C152 - Cholecystokinin (CCK) Fragment 26-33 Amide, Non-sulfated

EMAIL THIS PAGE TO A FRIEND
C152 Sigma

Cholecystokinin (CCK) Fragment 26-33 Amide, Non-sulfated

Synonym: CCK-8, non-sulfated

  • CAS Number 25679-24-7

  • Empirical Formula (Hill Notation) C49H62N10O13S2

  • Molecular Weight 1063.21

  •  MDL number MFCD00076488

Purchase

Properties

InChI Key   OIXQINQYMGNCII-UHFFFAOYSA-N
storage temp.   −20°C

Description

Amino Acid Sequence

Asp-Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C2901 Cholecystokinin (CCK) Fragment 26-33 Amide, Non-sulfated, ≥95% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?