EMAIL THIS PAGE TO A FRIEND
C180 Sigma-Aldrich

Colchicine

Synonym: (S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide

  • CAS Number 64-86-8

  • Empirical Formula (Hill Notation) C22H25NO6

  • Molecular Weight 399.44

  •  Beilstein/REAXYS Number 2228813

  •  EC Number 200-598-5

  •  MDL number MFCD00078484

Purchase

Properties

InChI Key   IAKHMKGGTNLKSZ-INIZCTEOSA-N
mp   150-160 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGI
WGK Germany 
3
RTECS 
GH0700000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C9754 Colchicine, ≥95% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?