• USA Home
  • C2681 - Z-Val-Lys-Met 7-amido-4-methylcoumarin

EMAIL THIS PAGE TO A FRIEND
C2681 Sigma

Z-Val-Lys-Met 7-amido-4-methylcoumarin

protease substrate

Purchase

Properties

composition   Peptide content, ~75%
storage temp.   −20°C

Description

Substrates

Substrate for amyloid A4-generating enzymes of Alzheimer′s disease.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?