• USA Home
  • C3093 - Chloroethylclonidine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
C3093 Sigma-Aldrich

Chloroethylclonidine dihydrochloride

Synonym: 2-[2,6-Dichloro(N-β-chloroethyl-N-methyl)-4-aminomethyl]phenylimino-2-imidazolidine dihydrochloride, CEC dihydrochloride

  • CAS Number 70107-07-2

  • Empirical Formula (Hill Notation) C13H17Cl3N4 · 2HCl

  • Molecular Weight 408.58

  •  MDL number MFCD00055181

Purchase

Properties

InChI Key   CZSAXNDACBPMAB-UHFFFAOYSA-N

Description

Application

Irreversible α1b adrenergic alkylating agent

Biochem/physiol Actions

Irreversible α1B-adrenoceptor alkylating agent.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B003 Chloroethylclonidine dihydrochloride, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?