EMAIL THIS PAGE TO A FRIEND
C3251 Sigma-Aldrich

Z-Ala-Leu

  • CAS Number 24959-68-0

  • Empirical Formula (Hill Notation) C17H24N2O5

  • Molecular Weight 336.38

  •  MDL number MFCD00038299

  •  PubChem Substance ID 24892595

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   YRQTWFBFEXBEQX-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?