• USA Home
  • C3488 - Creatinine standard solution

EMAIL THIS PAGE TO A FRIEND
C3488 Supelco

Creatinine standard solution

0.03 mg/mL

Purchase

Properties

concentration   0.03 mg/mL
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Solution contains creatinine in 0.02 N HCl.

Safety & Documentation

Safety Information

RIDADR 
UN 1789 8/PG 3
WGK Germany 
-

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?