EMAIL THIS PAGE TO A FRIEND
C3663 Sigma-Aldrich

Z-D-Glu-OBzl

Synonym: N-Cbz-D-glutamic acid α-benzyl ester, Z-D-glutamic acid 1-benzyl ester

  • CAS Number 65706-99-2

  • Empirical Formula (Hill Notation) C20H21NO6

  • Molecular Weight 371.38

  •  Beilstein/REAXYS Number 5305848

  •  MDL number MFCD00069647

Purchase

Properties

InChI Key   VWHKODOUMSMUAF-QGZVFWFLSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

96125 Z-D-Glu-OBzl, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?