• USA Home
  • C3767 - γ-Carboxy-DL-glutamic acid

EMAIL THIS PAGE TO A FRIEND
C3767 Sigma-Aldrich

γ-Carboxy-DL-glutamic acid

Synonym: 3-Amino-1,1,3-propanetricarboxylic acid

  • CAS Number 56271-99-9

  • Linear Formula NH2CH(COOH)CH2CH(COOH)2

  • Molecular Weight 191.14

  •  Beilstein/REAXYS Number 2214048

  •  EC Number 260-087-8

  •  MDL number MFCD00065488

Purchase

Properties

storage temp.   2-8°C
SMILES string   NC(CC(C(O)=O)C(O)=O)C(O)=O
InChI   1S/C6H9NO6/c7-3(6(12)13)1-2(4(8)9)5(10)11/h2-3H,1,7H2,(H,8,9)(H,10,11)(H,12,13)
InChI key   UHBYWPGGCSDKFX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

21885 γ-Carboxy-DL-glutamic acid, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?