EMAIL THIS PAGE TO A FRIEND
C3803 Sigma-Aldrich

Z-Asp(OMe)-OH

Synonym: N-Cbz-L-aspartic acid β-methyl ester, Z-L-Aspartic acid 4-methyl ester

  • CAS Number 3160-47-2

  • Linear Formula CH3O2CCH2CH(NHCO2CH2C6H5)CO2H

  • Molecular Weight 281.26

  •  Beilstein/REAXYS Number 6279422

  •  MDL number MFCD00133585

Purchase

Properties

storage temp.   −20°C
SMILES string   COC(=O)C[C@H](NC(=O)OCc1ccccc1)C(O)=O
InChI   1S/C13H15NO6/c1-19-11(15)7-10(12(16)17)14-13(18)20-8-9-5-3-2-4-6-9/h2-6,10H,7-8H2,1H3,(H,14,18)(H,16,17)/t10-/m0/s1
InChI key   PHMBNDDHIBIDRQ-JTQLQIEISA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

96002 Z-Asp(OMe)-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?