• USA Home
  • C3937 - Calcineurin autoinhibitory fragment

EMAIL THIS PAGE TO A FRIEND
C3937 Sigma-Aldrich

Calcineurin autoinhibitory fragment

≥90% (HPLC)

Synonym: Ile-Thr-Ser-Phe-Glu-Glu-Ala-Lys-Gly-Leu-Asp-Arg-Ile-Asn-Glu-Arg-Met-Pro-Pro-Arg-Arg-Asp-Ala-Met-Pro

  • CAS Number 148067-21-4

  • Empirical Formula (Hill Notation) C124H205N39O39S2

  • Molecular Weight 2930.32

  •  MDL number MFCD00214597

  •  PubChem Substance ID 24892659

Purchase

Properties

InChI Key   JRQDGUSHUYLSHC-UHFFFAOYSA-N
assay   ≥90% (HPLC)
storage temp.   −20°C

Description

Application

Membrane-impermeable blocker of calcineurin.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?