EMAIL THIS PAGE TO A FRIEND
C4251 Sigma-Aldrich

Z-β-Ala-Val

  • CAS Number 61058-48-8

  • Empirical Formula (Hill Notation) C16H22N2O5

  • Molecular Weight 322.36

  •  MDL number MFCD00056145

  •  PubChem Substance ID 24892676

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   XFMVOOGGOCFBNM-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?