EMAIL THIS PAGE TO A FRIEND
C4751 Sigma-Aldrich

Z-Arg-OH

Synonym: Nα-Z-L-Arginine, N2-Carbobenzyloxy-L-arginine

  • CAS Number 1234-35-1

  • Linear Formula HN=C(NH2)NH(CH2)3CH(NHCOOCH2C6H5)COOH

  • Molecular Weight 308.33

  •  Beilstein/REAXYS Number 2169267

  •  EC Number 214-973-6

  •  MDL number MFCD00001762

Purchase

Properties

mp   171-174 °C (dec.) (lit.)
storage temp.   −20°C
SMILES string   NC(=N)NCCC[C@H](NC(=O)OCc1ccccc1)C(O)=O
InChI   1S/C14H20N4O4/c15-13(16)17-8-4-7-11(12(19)20)18-14(21)22-9-10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9H2,(H,18,21)(H,19,20)(H4,15,16,17)/t11-/m0/s1
InChI key   SJSSFUMSAFMFNM-NSHDSACASA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

95930 Z-Arg-OH, ≥96.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?