• USA Home
  • C5207 - Calcineurin substrate

EMAIL THIS PAGE TO A FRIEND
C5207 Sigma-Aldrich

Calcineurin substrate

≥97% (HPLC)

Purchase

Description

Amino Acid Sequence

Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser-Val-Ala-Ala-Glu

Packaging

1 mg in glass bottle

250 μg in glass bottle

Biochem/physiol Actions

Peptide from the regulatory RII subunit of cAMP-dependent protein kinase. Useful in assays for protein phosphatase-2B (calcineurin) activity.

Other Notes

Lyophilized from 0.1% TFA in H2O

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?