EMAIL THIS PAGE TO A FRIEND
C5252 Sigma-Aldrich

Z-Ser-OH

Synonym: Carbobenzyloxy-L-serine, Z-L-Serine

  • CAS Number 1145-80-8

  • Linear Formula HOCH2CH(NHCO2CH2C6H5)CO2H

  • Molecular Weight 239.22

  •  Beilstein/REAXYS Number 2058314

  •  EC Number 214-546-4

  •  MDL number MFCD00002662

Purchase

Properties

mp   116-119 °C (lit.)
storage temp.   −20°C
SMILES string   OC[C@H](NC(=O)OCc1ccccc1)C(O)=O
InChI   1S/C11H13NO5/c13-6-9(10(14)15)12-11(16)17-7-8-4-2-1-3-5-8/h1-5,9,13H,6-7H2,(H,12,16)(H,14,15)/t9-/m0/s1
InChI key   GNIDSOFZAKMQAO-VIFPVBQESA-N

Description

Application

Building block in peptide synthesis; Starting material for the synthesis of various α-amino acids via the β-lactone

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

860700 Z-Ser-OH, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?