• USA Home
  • C5525 - Cholesteryl methyl carbonate

EMAIL THIS PAGE TO A FRIEND
C5525 Sigma-Aldrich

Cholesteryl methyl carbonate

Synonym: 3β-Hydroxy-5-cholestene 3-methyl carbonate, 5-Cholesten-3β-ol 3-methyl carbonate

  • CAS Number 15507-52-5

  • Empirical Formula (Hill Notation) C29H48O3

  • Molecular Weight 444.69

  •  MDL number MFCD00021136

Purchase

Properties

storage temp.   −20°C
SMILES string   COC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)[C@@H](C)CCCC(C)C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?