EMAIL THIS PAGE TO A FRIEND
C5904 Sigma-Aldrich

(±)-β-Citronellol

~95%

Synonym: 3,7-Dimethyl-6-octen-1-ol

  • CAS Number 106-22-9

  • Linear Formula (CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH

  • Molecular Weight 156.27

  •  Beilstein/REAXYS Number 1721507

  •  EC Number 203-375-0

  •  MDL number MFCD00002935

Purchase

Properties

vapor density   5.4 (vs air)
vapor pressure   ~0.02 mmHg ( 25 °C)
InChI Key   QMVPMAAFGQKVCJ-UHFFFAOYSA-N
assay   ~95%
refractive index   n20/D 1.456 (lit.)
bp   222 °C(lit.)
density   0.857 g/mL at 25 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
210.2 °F
Flash Point(C) 
99 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C83201 β-Citronellol, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?