EMAIL THIS PAGE TO A FRIEND
C6626 Sigma-Aldrich

Z-Cys(Z)-OH

Synonym: N,S-Di-Z-L-cysteine

  • CAS Number 57912-35-3

  • Empirical Formula (Hill Notation) C19H19NO6S

  • Molecular Weight 389.42

  •  Beilstein/REAXYS Number 2547850

  •  EC Number 261-013-7

  •  MDL number MFCD00037823

Purchase

Properties

InChI Key   PXKPRICKEUGRRR-INIZCTEOSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?